siRNA Details
| Gene Symbol: ERGIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016570.2 | NP_057654.2 | 5 | 412 | ||
| GenBank mRNA | AF183410.1 | - | 5 | 363 | ||
| AF216751.1 | - | 5 | 367 | |||
| AF302767.1 | - | 5 | 365 | |||
| AK074520.1 | - | 5 | 387 | |||
| AK222799.1 | - | 5 | 352 | |||
| AL834128.2 | - | 5 | 353 | |||
| BC000887.1 | - | 5 | 365 | 3462408 | ||
| BC107794.1 | - | 5 | 369 | 30319807 | ||
| BI547753.1 | - | 5 | 416 | |||

ERGIC2
Genomic Map