siRNA Details
| Gene Symbol: RNPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017619.3 | NP_060089.1 | 10 | 1228 | ||
| GenBank mRNA | AK000015.1 | - | 4 | 403 | ||
| AK057799.1 | - | 10 | 1207 | |||
| AK289844.1 | - | 10 | 1490 | |||
| AL137730.1 | - | 4 | 440 | |||
| AY099329.1 | - | 10 | 1213 | |||
| BC010697.1 | - | 10 | 1204 | 3873751 | ||

RNPC3
Genomic Map