siRNA Details
| Gene Symbol: ASAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143778.1 | NP_001137250.1 | 9 | 994 | ||
| NM_017707.3 | NP_060177.2 | 10 | 1021 | |||
| XM_011541755.1 | XP_011540057.1 | 10 | 961 | |||
| XM_017001685.1 | XP_016857174.1 | 10 | 955 | |||
| XM_017001686.1 | XP_016857175.1 | 10 | 961 | |||
| XM_017001687.1 | XP_016857176.1 | 10 | 928 | |||
| XM_017001688.1 | XP_016857177.1 | 10 | 917 | |||
| XM_017001689.1 | XP_016857178.1 | 10 | 978 | |||
| GenBank mRNA | AB056749.1 | - | 10 | 942 | ||
| AK092291.1 | - | 10 | 943 | |||
| AK295100.1 | - | 9 | 916 | |||
| AK299359.1 | - | 10 | 917 | |||
| BC060786.1 | - | 10 | 969 | 5298355 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001008232, BC085250, AK132294.

ASAP3
Genomic Map