siRNA Details
Gene Symbol: FAM46C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017709.3 | NP_060179.2 | 2 | 4983 | ||
GenBank mRNA | AK091327.1 | - | 1 | 2166 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM46C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017709.3 | NP_060179.2 | 2 | 4983 | ||
GenBank mRNA | AK091327.1 | - | 1 | 2166 |