siRNA Details
Gene Symbol: TTC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017904.3 | NP_060374.2 | 6 | 1296 | ||
GenBank mRNA | AK000626.1 | - | 6 | 1226 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TTC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017904.3 | NP_060374.2 | 6 | 1296 | ||
GenBank mRNA | AK000626.1 | - | 6 | 1226 |