siRNA Details
| Gene Symbol: ZNHIT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017953.3 | NP_060423.3 | 1 | 397 | ||
| XM_011541614.2 | XP_011539916.1 | 1 | 411 | |||
| GenBank mRNA | AF308296.1 | - | 1 | 44 | ||
| AK000767.1 | - | 1 | 306 | |||
| AK314566.1 | - | 1 | 325 | |||
| AL442074.1 | - | 1 | 382 | |||
| BC026236.1 | - | 1 | 384 | 4664103 | ||
| BC110898.1 | - | 1 | 315 | 6012059 | ||
| DB169943.1 | - | 1 | 397 | |||

ZNHIT6
Genomic Map