siRNA Details
| Gene Symbol: GPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018066.3 | NP_060536.3 | 2 | 833 | ||
| GenBank mRNA | AK001211.1 | - | 2 | 752 | ||
| AK022535.1 | - | 2 | 738 | |||
| AY358864.1 | - | 2 | 742 | |||
| AY364261.1 | - | 2 | 551 | |||
| BC007815.2 | - | 2 | 675 | 4299794 | ||
| BC008634.1 | - | 2 | 699 | 3463813 | ||
| BC020272.1 | - | 2 | 718 | |||

GPN2
Genomic Map