siRNA Details
| Gene Symbol: NDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168551.1 | NP_001162023.1 | 5 | 699 | ||
| NM_018087.4 | NP_060557.3 | 5 | 819 | |||
| NR_033142.1 | 4 | 698 | ||||
| XM_011541766.2 | XP_011540068.1 | 5 | 661 | |||
| GenBank mRNA | AK001269.1 | - | 5 | 702 | ||
| AK295000.1 | - | 5 | 483 | |||
| AK298909.1 | - | 4 | 442 | |||
| AK302910.1 | - | 4 | 413 | |||
| AL354612.1 | - | 5 | 702 | |||
| AL354613.1 | - | 5 | 699 | |||
| BC003082.2 | - | 5 | 769 | 3509727 | ||
| BX648031.1 | - | 4 | 429 | |||
| DQ141696.1 | - | 5 | 528 | |||

NDC1
Genomic Map