siRNA Details
| Gene Symbol: GOLPH3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018178.5 | NP_060648.2 | 5 | 2499 | ||
| XM_006711428.1 | XP_006711491.1 | 5 | 2426 | |||
| GenBank mRNA | AJ296153.1 | - | 5 | 2325 | ||
| AK001549.1 | - | 4 | 2125 | |||
| BC013870.2 | - | 4 | 2298 | 3837466 | ||

GOLPH3L
Genomic Map