siRNA Details
| Gene Symbol: ETNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297760.1 | NP_001284689.1 | 8 | 2186 | ||
| NM_001297761.1 | NP_001284690.1 | 7 | 1820 | |||
| NM_001297762.1 | NP_001284691.1 | 7 | 1957 | |||
| NM_018208.3 | NP_060678.2 | 8 | 2080 | |||
| XM_011509714.2 | XP_011508016.1 | 7 | 1426 | |||
| XM_011509715.1 | XP_011508017.1 | 7 | 1731 | |||
| XM_017001632.1 | XP_016857121.1 | 7 | 1514 | |||
| GenBank mRNA | AB073608.1 | - | 8 | 1680 | ||
| AK001623.1 | - | 8 | 1898 | |||
| AK316493.1 | - | 7 | 1316 | |||
| BC010082.2 | - | 8 | 1901 | 4329734 | ||

ETNK2
Genomic Map