siRNA Details
| Gene Symbol: RSBN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018364.4 | NP_060834.2 | 1 | 698 | ||
| NR_130896.1 | 1 | 698 | ||||
| XM_017001518.1 | XP_016857007.1 | 1 | 698 | |||
| GenBank mRNA | AK002082.1 | - | 1 | 513 | ||
| AK292552.1 | - | 1 | 698 | |||
| BC026155.1 | - | 1 | 520 | 4827655 | ||
| CR627402.1 | - | 2 | 291 | |||

RSBN1
Genomic Map