siRNA Details
| Gene Symbol: RSBN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018364.4 | NP_060834.2 | 3 | 1486 | ||
| NR_130896.1 | 4 | 1604 | ||||
| GenBank mRNA | AK002082.1 | - | 3 | 1301 | ||
| AK022166.1 | - | 2 | 665 | |||
| AK292552.1 | - | 3 | 1486 | |||
| BC026155.1 | - | 4 | 1426 | 4827655 | ||
| CR627402.1 | - | 4 | 1079 | |||

RSBN1
Genomic Map