siRNA Details
| Gene Symbol: STYK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018423.2 | NP_060893.2 | 6 | 1090 | ||
| XM_005253417.2 | XP_005253474.1 | 5 | 990 | |||
| XM_011520736.1 | XP_011519038.1 | 7 | 893 | |||
| XM_011520737.2 | XP_011519039.1 | 6 | 930 | |||
| XM_011520738.2 | XP_011519040.1 | 6 | 1085 | |||
| XM_017019579.1 | XP_016875068.1 | 6 | 873 | |||
| GenBank mRNA | AK313906.1 | - | 6 | 825 | ||
| AL353940.1 | - | 6 | 838 | |||
| BC093822.1 | - | 4 | 591 | 7939667 | ||
| BC093824.1 | - | 4 | 591 | 7939669 | ||
| KP729000.1 | - | 4 | 569 | |||

STYK1
Genomic Map