siRNA Details
| Gene Symbol: CTTNBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018704.2 | NP_061174.1 | 6 | 2177 | ||
| XM_011541781.2 | XP_011540083.1 | 6 | 2248 | |||
| XM_017001806.1 | XP_016857295.1 | 6 | 2615 | |||
| GenBank mRNA | AK022544.1 | - | 5 | 2098 | ||
| BC016029.1 | - | 6 | 2117 | 3921313 | ||

CTTNBP2NL
Genomic Map