siRNA Details
| Gene Symbol: ELMOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130037.1 | NP_001123509.1 | 9 | 1068 | ||
| NM_001308018.1 | NP_001294947.1 | 11 | 1228 | |||
| NM_018712.3 | NP_061182.3 | 10 | 1092 | |||
| XM_017017994.1 | XP_016873483.1 | 10 | 1199 | |||
| XM_017017995.1 | XP_016873484.1 | 9 | 949 | |||
| GenBank mRNA | AK054794.1 | - | 9 | 888 | ||
| AK297351.1 | - | 11 | 1228 | |||
| AK303689.1 | - | 9 | 910 | |||
| AL359601.1 | - | 4 | 279 | |||
| BC028725.2 | - | 10 | 835 | 4815682 | ||

ELMOD1
Genomic Map