siRNA Details
| Gene Symbol: C11orf71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271562.1 | NP_001258491.1 | 1 | 522 | ||
| NM_019021.3 | NP_061894.2 | 1 | 522 | |||
| GenBank mRNA | AB101209.1 | - | 1 | 511 | ||
| AK000017.1 | - | 1 | 483 | |||
| BC009041.1 | - | 1 | 451 | 3868744 | ||
| BC071695.1 | - | 1 | 389 | 5416335 | ||
| CR457222.1 | - | 1 | 303 | |||

C11orf71
Genomic Map