siRNA Details
| Gene Symbol: FAM212B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019099.4 | NP_061972.1 | 2 | 1046 | ||
| NM_198926.2 | NP_945120.1 | 2 | 904 | |||
| XM_011541783.2 | XP_011540085.1 | 4 | 1142 | |||
| GenBank mRNA | AK055667.1 | - | 2 | 1022 | ||
| AK293989.1 | - | 2 | 931 | |||
| AL137198.1 | - | 2 | 953 | |||
| AL137199.1 | - | 2 | 904 | |||
| BC031558.1 | - | 2 | 1001 | 5181834 | ||

FAM212B
Genomic Map