siRNA Details
| Gene Symbol: FAM212B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019099.4 | NP_061972.1 | 2 | 1880 | ||
| NM_198926.2 | NP_945120.1 | 2 | 1738 | |||
| XM_011541783.2 | XP_011540085.1 | 4 | 1976 | |||
| GenBank mRNA | AK055667.1 | - | 2 | 1856 | ||

FAM212B
Genomic Map