siRNA Details
| Gene Symbol: CRTAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304782.1 | NP_001291711.1 | 2, 3 | 329 | ||
| NM_019604.3 | NP_062550.2 | 7, 8 | 877 | |||
| XM_011542900.2 | XP_011541202.1 | 6, 7 | 803 | |||
| GenBank mRNA | AB209830.1 | - | 2, 3 | 238 | ||
| AF001622.1 | - | 7, 8 | 828 | |||
| AK293470.1 | - | 2, 3 | 258 | |||
| AK313578.1 | - | 7, 8 | 875 | |||
| BC070266.1 | - | 7, 8 | 877 | 30418882 | ||
| BX118379.1 | - | 1, 2 | 44 | |||
| DC305734.1 | - | 2, 3 | 329 | |||

CRTAM
Genomic Map