siRNA Details
| Gene Symbol: TMEM167B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322248.1 | NP_001309177.1 | 3 | 2780 | ||
| NM_020141.3 | NP_064526.1 | 3 | 2679 | |||
| NR_136242.1 | 4 | 2808 | ||||
| NR_136243.1 | 2 | 2587 | ||||
| GenBank mRNA | AK075191.1 | - | 3 | 2701 | ||
| BC008739.2 | - | 3 | 2679 | 3346340 | ||
| BC065186.1 | - | 3 | 2679 | 4642864 | ||
| BC066918.1 | - | 3 | 2708 | 4826492 | ||
| BC067116.1 | - | 3 | 2665 | 5532892 | ||

TMEM167B
Genomic Map