siRNA Details
| Gene Symbol: COQ9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020312.3 | NP_064708.1 | 7 | 869 | ||
| GenBank mRNA | AF161444.1 | - | 7 | 803 | ||
| AK290627.1 | - | 7 | 799 | |||
| AL136884.1 | - | 2 | 303 | |||
| BC001478.2 | - | 5 | 444 | |||
| BC029341.1 | - | 7 | 803 | |||
| BC054340.1 | - | 7 | 803 | 6514554 | ||
| BC064946.1 | - | 7 | 804 | 6023461 | ||

COQ9
Genomic Map