siRNA Details
| Gene Symbol: COQ9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020312.3 | NP_064708.1 | 9 | 1201 | ||
| GenBank mRNA | AF161444.1 | - | 9 | 1135 | ||
| AL136884.1 | - | 4 | 635 | |||
| BC001478.2 | - | 7 | 776 | |||
| BC029341.1 | - | 9 | 1135 | |||
| BC054340.1 | - | 9 | 1135 | 6514554 | ||
| BC064946.1 | - | 9 | 1136 | 6023461 | ||

COQ9
Genomic Map