siRNA Details
| Gene Symbol: ZNF687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304763.1 | NP_001291692.1 | 4 | 3211 | ||
| NM_001304764.1 | NP_001291693.1 | 3 | 2412 | |||
| NM_020832.2 | NP_065883.1 | 3 | 2342 | |||
| XM_005245366.3 | XP_005245423.1 | 3 | 2780 | |||
| XM_011509811.2 | XP_011508113.1 | 3 | 2389 | |||
| XM_011509812.2 | XP_011508114.1 | 4 | 2659 | |||
| XM_011509813.2 | XP_011508115.1 | 3 | 2543 | |||
| GenBank mRNA | AB037862.1 | - | 4 | 3211 | ||
| AK023105.1 | - | 2 | 1049 | |||
| BC032463.1 | - | 3 | 2339 | 5196247 | ||
| CR749307.1 | - | 3 | 2412 | |||

ZNF687
Genomic Map