siRNA Details
| Gene Symbol: PRUNE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303229.1 | NP_001290158.1 | 7 | 2107 | ||
| NM_001303242.1 | NP_001290171.1 | 7 | 2151 | |||
| NM_001303243.1 | NP_001290172.1 | 6 | 1966 | |||
| NM_021222.2 | NP_067045.1 | 8 | 2310 | |||
| NR_130130.1 | 4 | 1670 | ||||
| NR_130131.1 | 5 | 1763 | ||||
| NR_130132.1 | 4 | 1604 | ||||
| NR_130135.1 | 5 | 1763 | ||||
| XM_005245393.4 | XP_005245450.1 | 8 | 2198 | |||
| XM_011509832.2 | XP_011508134.1 | 6 | 1999 | |||
| XM_017001955.1 | XP_016857444.1 | 7 | 2039 | |||
| XM_017001956.1 | XP_016857445.1 | 6 | 1908 | |||
| XM_017001957.1 | XP_016857446.1 | 6 | 1855 | |||
| GenBank mRNA | AF051907.1 | - | 8 | 2224 | ||
| AF123538.1 | - | 4 | 1584 | |||
| AF123539.1 | - | 7 | 1991 | |||
| AK027875.1 | - | 4 | 1443 | |||
| AK298273.1 | - | 4 | 1438 | |||
| AL122054.1 | - | 1 | 968 | |||
| AL137324.1 | - | 1 | 591 | |||
| BC014886.1 | - | 7 | 2015 | 3907577 | ||
| BC017755.1 | - | 1 | 473 | |||
| BC025304.1 | - | 8 | 2204 | 5458046 | ||
| BC063481.1 | - | 4 | 1465 | |||

PRUNE1
Genomic Map