siRNA Details
| Gene Symbol: GREM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022469.3 | NP_071914.3 | 2 | 1208 | ||
| XM_005273226.3 | XP_005273283.1 | 2 | 1036 | |||
| XM_011544249.2 | XP_011542551.1 | 3 | 1538 | |||
| GenBank mRNA | AK024848.1 | - | 2 | 1195 | ||
| BC046632.1 | - | 2 | 1135 | 5288555 | ||

GREM2
Genomic Map