siRNA Details
| Gene Symbol: RSRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023012.5 | NP_075388.2 | 2 | 243 | ||
| NR_036434.1 | 2 | 243 | ||||
| NR_036435.1 | 2 | 243 | ||||
| NR_036436.1 | 2 | 243 | ||||
| XM_005253601.1 | XP_005253658.1 | 2 | 243 | |||
| XM_005253602.2 | XP_005253659.1 | 2 | 192 | |||
| XM_005253604.1 | XP_005253661.1 | 2 | 225 | |||
| XM_011538688.1 | XP_011536990.1 | 2 | 243 | |||
| XM_011538689.1 | XP_011536991.1 | 4 | 398 | |||
| XM_017019830.1 | XP_016875319.1 | 2 | 243 | |||
| XM_017019831.1 | XP_016875320.1 | 3 | 298 | |||
| XM_017019832.1 | XP_016875321.1 | 4 | 398 | |||
| XM_017019833.1 | XP_016875322.1 | 2 | 192 | |||
| GenBank mRNA | AB212664.1 | - | 2 | 229 | ||
| AK001883.1 | - | 2 | 111 | |||
| AK056114.1 | - | 2 | 166 | |||
| AK056256.1 | - | 2 | 200 | |||
| BC008684.1 | - | 2 | 153 | 3853928 | ||
| BC067773.1 | - | 2 | 213 | 5301415 | ||
| BX640711.1 | - | 2 | 167 | |||
| BX647884.1 | - | 2 | 210 | |||
| DB081070.1 | - | 2 | 244 | |||

RSRC2
Genomic Map