siRNA Details
| Gene Symbol: C1orf116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083924.1 | NP_001077393.1 | 3 | 1117 | ||
| NM_023938.5 | NP_076427.2 | 4 | 1303 | |||
| XM_005273259.1 | XP_005273316.1 | 2 | 871 | |||
| XM_006711530.1 | XP_006711593.1 | 4 | 1180 | |||
| XM_011509973.2 | XP_011508275.1 | 3 | 973 | |||
| GenBank mRNA | AK093826.1 | - | 4 | 1271 | ||
| AL832940.1 | - | 1 | 377 | |||
| AY352640.1 | - | 4 | 1294 | |||
| BC000765.2 | - | 4 | 1199 | 2822914 | ||
| BC018959.2 | - | 2 | 960 | |||

C1orf116
Genomic Map