siRNA Details
| Gene Symbol: C1orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024097.3 | NP_077002.2 | 3, 4 | 357 | ||
| NR_040733.1 | 2, 3 | 241 | ||||
| GenBank mRNA | BC001508.2 | - | 3, 4 | 333 | 2959109 | |
| BC001711.1 | - | 3, 4 | 333 | 3543543 | ||
| BM702424.1 | - | 2, 3 | 193 | |||
| CN256666.1 | - | 2, 3 | 141 | |||

C1orf50
Genomic Map