siRNA Details
| Gene Symbol: PTMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002824.4 | NP_002815.3 | 4, 5 | 597 | ||
| XM_011520987.1 | XP_011519289.1 | 4, 5 | 910 | |||
| GenBank mRNA | BC017025.1 | - | 4, 5 | 316 | 3900998 | |
| BC128229.1 | - | 5, 6 | 264 | |||
| BE791815.1 | - | 1, 2 | 47 | |||
| BM550529.1 | - | 674 | ||||
| M24398.1 | - | 4, 5 | 566 | |||

PTMS
Genomic Map