siRNA Details
| Gene Symbol: C11orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302084.1 | NP_001289013.1 | 3 | 307 | ||
| NM_024650.3 | NP_078926.3 | 5 | 599 | |||
| GenBank mRNA | AK026184.1 | - | 4 | 400 | ||
| AK225435.1 | - | 4 | 400 | |||
| AK302025.1 | - | 3 | 259 | |||
| BC028240.1 | - | 5 | 602 | 5243907 | ||
| BI559574.1 | - | 5 | 515 | |||

C11orf80
Genomic Map