siRNA Details
| Gene Symbol: C11orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024650.3 | NP_078926.3 | 8, 9 | 849 | ||
| GenBank mRNA | AK026184.1 | - | 7, 8 | 650 | ||
| AK225435.1 | - | 7, 8 | 650 | |||
| BC028240.1 | - | 8, 9 | 852 | 5243907 | ||

C11orf80
Genomic Map