siRNA Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024667.2 | NP_078943.1 | 2 | 314 | ||
| XM_005253622.3 | XP_005253679.1 | 3 | 3029 | |||
| XM_006719602.3 | XP_006719665.1 | 3 | 350 | |||
| XM_011538743.2 | XP_011537045.1 | 3 | 455 | |||
| XM_011538745.2 | XP_011537047.1 | 2 | 2845 | |||
| XM_017019970.1 | XP_016875459.1 | 3 | 459 | |||
| GenBank mRNA | AK022812.1 | - | 2 | 314 | ||
| BC005882.2 | - | 2 | 252 | 3533384 | ||

VPS37B
Genomic Map