siRNA Details
| Gene Symbol: C1orf115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024709.4 | NP_078985.3 | 2 | 1811 | ||
| GenBank mRNA | AK024208.1 | - | 2 | 1732 | ||
| AK056749.1 | - | 3 | 1677 | |||
| AK091937.1 | - | 2 | 1745 | |||
| AK125403.1 | - | 2 | 2251 | |||
| BC036067.1 | - | 2 | 1777 | 5274863 | ||

C1orf115
Genomic Map