siRNA Details
| Gene Symbol: CCDC82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318736.1 | NP_001305665.1 | 5 | 756 | ||
| NM_001318737.1 | NP_001305666.1 | 4 | 621 | |||
| NM_024725.3 | NP_079001.2 | 4 | 621 | |||
| XM_005274292.3 | XP_005274349.1 | 2 | 501 | |||
| XM_006718911.3 | XP_006718974.1 | 5 | 642 | |||
| XM_011542984.2 | XP_011541286.1 | 4 | 1285 | |||
| XM_011542987.2 | XP_011541289.1 | 5 | 717 | |||
| XM_011542988.2 | XP_011541290.1 | 3 | 593 | |||
| XM_017018305.1 | XP_016873794.1 | 3 | 2366 | |||
| XM_017018306.1 | XP_016873795.1 | 4 | 2450 | |||
| XM_017018307.1 | XP_016873796.1 | 5 | 1092 | |||
| XM_017018308.1 | XP_016873797.1 | 2 | 2326 | |||
| XM_017018309.1 | XP_016873798.1 | 2 | 461 | |||
| XM_017018310.1 | XP_016873799.1 | 2 | 502 | |||
| XM_017018311.1 | XP_016873800.1 | 4 | 2366 | |||
| XM_017018312.1 | XP_016873801.1 | 5 | 1285 | |||
| GenBank mRNA | AF245436.1 | - | 4 | 609 | ||
| AK056790.1 | - | 4 | 614 | |||
| AK074306.1 | - | 5 | 750 | |||
| AK314669.1 | - | 1 | 650 | |||
| BC018663.2 | - | 3 | 556 | 2964737 | ||
| BC033726.2 | - | 1 | 422 | 5001231 | ||

CCDC82
Genomic Map