siRNA Details
| Gene Symbol: VASH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024749.4 | NP_079025.2 | 3, 4 | 792 | ||
| GenBank mRNA | BC051856.1 | - | 3, 4 | 677 | 6067012 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

VASH2| Gene Symbol: VASH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024749.4 | NP_079025.2 | 3, 4 | 792 | ||
| GenBank mRNA | BC051856.1 | - | 3, 4 | 677 | 6067012 | |