siRNA Details
| Gene Symbol: QSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076786.2 | NP_001070254.1 | 4 | 586 | ||
| XM_006718325.3 | XP_006718388.3 | 4 | 938 | |||
| XM_017018327.1 | XP_016873816.1 | 4 | 938 | |||
| XM_017018328.1 | XP_016873817.1 | 4 | 938 | |||
| XM_017018329.1 | XP_016873818.1 | 2 | 1391 | |||
| XM_017018330.1 | XP_016873819.1 | 3 | 336 | |||
| GenBank mRNA | AK126023.1 | - | 4 | 607 | ||
| BC112935.1 | - | 4 | 586 | |||

QSER1
Genomic Map