siRNA Details
| Gene Symbol: ZNF669 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142572.1 | NP_001136044.1 | 4 | 1060 | ||
| NM_024804.2 | NP_079080.2 | 4 | 1318 | |||
| XM_005273270.2 | XP_005273327.1 | 3 | 1027 | |||
| XM_005273271.2 | XP_005273328.1 | 3 | 1285 | |||
| GenBank mRNA | AK022668.1 | - | 4 | 1272 | ||
| AK056020.1 | - | 4 | 1049 | |||
| BC015312.1 | - | 4 | 1318 | 4413005 | ||

ZNF669
Genomic Map