siRNA Details
| Gene Symbol: PYGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002862.3 | NP_002853.2 | 12 | 1527 | ||
| GenBank mRNA | AB209080.1 | - | 12 | 1485 | ||
| AK299697.1 | - | 12 | 1451 | |||
| AK309036.1 | - | 12 | 1525 | |||
| BC017045.2 | - | 12 | 1502 | 3877951 | ||
| BC030795.2 | - | 12 | 1503 | 3916938 | ||
| J03544.1 | - | 12 | 1497 | |||
| U47025.1 | - | 12 | 1452 | |||

PYGB
Genomic Map