siRNA Details
| Gene Symbol: ZNF672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024836.2 | NP_079112.1 | 4 | 3047 | ||
| XM_005270336.1 | XP_005270393.1 | 4 | 2892 | |||
| GenBank mRNA | AK025954.1 | - | 4 | 2894 | ||
| AK027476.1 | - | 1 | 2072 | |||
| AK027679.1 | - | 2 | 2441 | |||
| BC007385.2 | - | 1 | 1501 | |||
| BC008872.1 | - | 5 | 1533 | |||
| BC035140.1 | - | 1 | 3427 | 5264126 | ||
| BC050348.1 | - | 1 | 3423 | 5260682 | ||
| BC068506.1 | - | 4 | 2745 | 4824147 | ||

ZNF672
Genomic Map