siRNA Details
| Gene Symbol: TARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271895.1 | NP_001258824.1 | 4 | 555 | ||
| NM_001271896.1 | NP_001258825.1 | 4 | 555 | |||
| NM_025150.4 | NP_079426.2 | 4 | 555 | |||
| XM_006711555.1 | XP_006711618.1 | 4 | 518 | |||
| XM_006711556.1 | XP_006711619.1 | 4 | 518 | |||
| XM_017002394.1 | XP_016857883.1 | 4 | 518 | |||
| XM_017002395.1 | XP_016857884.1 | 4 | 518 | |||
| GenBank mRNA | AK022590.1 | - | 4 | 505 | ||
| AK222814.1 | - | 4 | 496 | |||
| AK225368.1 | - | 4 | 475 | |||
| AK315507.1 | - | 4 | 501 | |||
| AY450558.1 | - | 3, 4 | 313 | |||
| BC000541.2 | - | 4 | 502 | 3163506 | ||
| BC007824.2 | - | 4 | 443 | |||
| BC009997.2 | - | 4 | 475 | 4128881 | ||

TARS2
Genomic Map