siRNA Details
| Gene Symbol: PAAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267803.1 | NP_001254732.1 | 4 | 287 | ||
| NM_001267804.1 | NP_001254733.1 | 4 | 278 | |||
| NM_001267805.1 | NP_001254734.1 | 4 | 274 | |||
| NM_001267806.1 | NP_001254735.1 | 3 | 161 | |||
| NM_025155.2 | NP_079431.1 | 3 | 218 | |||
| XM_005274309.1 | XP_005274366.1 | 2 | 118 | |||
| XM_017018357.1 | XP_016873846.1 | 4 | 247 | |||
| XM_017018358.1 | XP_016873847.1 | 4 | 252 | |||
| GenBank mRNA | AF087895.1 | - | 4 | 212 | ||
| AK021910.1 | - | 3 | 199 | |||
| AK222501.1 | - | 3 | 199 | |||
| AK225520.1 | - | 3 | 186 | |||
| AK298258.1 | - | 3 | 161 | |||
| AK316394.1 | - | 3 | 432 | |||
| BC021541.1 | - | 3 | 182 | 3942680 | ||
| BC028628.1 | - | 4 | 252 | 5263875 | ||
| CD243380.1 | - | 4 | 264 | |||
| DC354686.1 | - | 3 | 220 | |||

PAAF1
Genomic Map