siRNA Details
| Gene Symbol: ANKRD13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030816.4 | NP_110443.3 | 4, 5 | 997 | ||
| XM_005271234.3 | XP_005271291.1 | 4, 5 | 1240 | |||
| XM_005271235.3 | XP_005271292.1 | 4, 5 | 987 | |||
| XM_006710929.3 | XP_006710992.1 | 4, 5 | 1010 | |||
| XM_017002413.1 | XP_016857902.1 | 2, 3 | 220 | |||
| XM_017002415.1 | XP_016857904.1 | 4, 5 | 348 | |||
| GenBank mRNA | AL136717.1 | - | 3, 4 | 893 | ||
| BC028840.1 | - | 4, 5 | 986 | 4558748 | ||

ANKRD13C
Genomic Map