siRNA Details
| Gene Symbol: ISG20L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303095.1 | NP_001290024.1 | 2 | 283 | ||
| XM_005245516.4 | XP_005245573.1 | 2 | 251 | |||
| XM_006711560.3 | XP_006711623.1 | 2 | 421 | |||
| XM_006711561.3 | XP_006711624.1 | 2 | 310 | |||
| GenBank mRNA | AK022733.1 | - | 1 | 341 | ||
| AK095697.1 | - | 1 | 863 | |||
| AY391837.1 | - | 1 | 269 | |||
| BC000575.2 | - | 2 | 263 | 3162193 | ||
| HY144219.1 | - | 2 | 284 | |||

ISG20L2
Genomic Map