siRNA Details
| Gene Symbol: DDX59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031725.5 | NP_001026895.2 | 2 | 344 | ||
| NM_001320181.1 | NP_001307110.1 | 2 | 344 | |||
| NM_001320182.1 | NP_001307111.1 | 1 | 112 | |||
| XM_005245519.1 | XP_005245576.1 | 2 | 253 | |||
| XM_005245521.2 | XP_005245578.1 | 2 | 253 | |||
| XM_006711562.1 | XP_006711625.1 | 2 | 253 | |||
| XM_011510035.2 | XP_011508337.1 | 2 | 277 | |||
| XM_017002431.1 | XP_016857920.1 | 2 | 1032 | |||
| XM_017002432.1 | XP_016857921.1 | 2 | 517 | |||
| GenBank mRNA | AK299219.1 | - | 2 | 215 | ||
| AK307569.1 | - | 272 | ||||
| AL136611.1 | - | 2 | 257 | |||
| BC014183.2 | - | 2 | 277 | |||
| BC041801.1 | - | 2 | 335 | 5302098 | ||

DDX59
Genomic Map