siRNA Details
| Gene Symbol: STK40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282546.1 | NP_001269475.1 | 11 | 2070 | ||
| NM_001282547.1 | NP_001269476.1 | 11 | 2055 | |||
| NM_032017.2 | NP_114406.1 | 12 | 2292 | |||
| GenBank mRNA | AK024504.1 | - | 9 | 3244 | ||
| AK075029.1 | - | 8 | 1571 | |||
| AK075048.1 | - | 11 | 2049 | |||
| AK131560.1 | - | 3 | 2572 | |||
| AL834137.1 | - | 12 | 2249 | |||
| AY198395.1 | - | 11 | 2025 | |||
| BC005169.2 | - | 6 | 1254 | |||
| BC007835.2 | - | 11 | 2023 | 4109652 | ||
| BC008344.1 | - | 11 | 2031 | 3530483 | ||

STK40
Genomic Map