siRNA Details
| Gene Symbol: FAM160A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL136723.1 | - | 9 | 1838 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM160A2| Gene Symbol: FAM160A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL136723.1 | - | 9 | 1838 | ||