siRNA Details
| Gene Symbol: LINC00467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026761.2 | 3 | 308 | |||
| GenBank mRNA | AK130857.1 | - | 3 | 275 | ||
| AK130858.1 | - | 3 | 260 | |||
| BC005997.1 | - | 3 | 244 | |||
| BX382054.2 | - | 3 | 243 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00467| Gene Symbol: LINC00467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026761.2 | 3 | 308 | |||
| GenBank mRNA | AK130857.1 | - | 3 | 275 | ||
| AK130858.1 | - | 3 | 260 | |||
| BC005997.1 | - | 3 | 244 | |||
| BX382054.2 | - | 3 | 243 | |||