siRNA Details
| Gene Symbol: MFSD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136493.2 | NP_001129965.1 | 6 | 850 | ||
| NM_001287808.1 | NP_001274737.1 | 4 | 551 | |||
| NM_001287809.1 | NP_001274738.1 | 6 | 700 | |||
| NM_032793.4 | NP_116182.2 | 6 | 811 | |||
| NR_109896.1 | 6 | 811 | ||||
| XM_005271285.1 | XP_005271342.1 | 6 | 775 | |||
| XM_011542312.2 | XP_011540614.1 | 6 | 778 | |||
| GenBank mRNA | AB073383.1 | - | 6 | 775 | ||
| AF289609.1 | - | 6 | 767 | |||
| AF370364.1 | - | 3 | 1276 | |||
| AK074186.1 | - | 6 | 716 | |||
| AK075183.1 | - | 6 | 778 | |||
| AK093223.1 | - | 6 | 812 | |||
| AK291540.1 | - | 6 | 773 | |||
| AK297991.1 | - | 6 | 664 | |||
| AK301533.1 | - | 4 | 518 | |||
| AL834184.1 | - | 6 | 765 | |||
| AY358636.1 | - | 6 | 793 | |||
| BC006353.2 | - | 2 | 146 | |||
| BC011587.1 | - | 6 | 785 | 4646430 | ||
| BC092414.1 | - | 6 | 753 | 30387073 | ||

MFSD2A
Genomic Map