siRNA Details
| Gene Symbol: C1orf198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136494.1 | NP_001129966.1 | 6 | 3132 | ||
| NM_001136495.1 | NP_001129967.1 | 4 | 2856 | |||
| NM_032800.2 | NP_116189.1 | 4 | 3074 | |||
| XM_017002599.1 | XP_016858088.1 | 5 | 3339 | |||
| GenBank mRNA | AK027431.1 | - | 4 | 3047 | ||
| AK131306.1 | - | 4 | 2982 | |||
| AL713769.1 | - | 4 | 2822 | |||
| AL833809.1 | - | 1 | 316 | |||
| BC004870.2 | - | 1 | 1829 | |||
| BC066649.1 | - | 4 | 3064 | 6011914 | ||

C1orf198
Genomic Map