siRNA Details
| Gene Symbol: C1orf198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136494.1 | NP_001129966.1 | 6 | 3521 | ||
| NM_001136495.1 | NP_001129967.1 | 4 | 3245 | |||
| NM_032800.2 | NP_116189.1 | 4 | 3463 | |||
| XM_017002599.1 | XP_016858088.1 | 5 | 3728 | |||
| GenBank mRNA | AK027431.1 | - | 4 | 3436 | ||
| AK131306.1 | - | 4 | 3371 | |||
| AL713769.1 | - | 4 | 3211 | |||
| AL833809.1 | - | 1 | 705 | |||
| BC004870.2 | - | 1 | 2218 | |||
| BC066649.1 | - | 4 | 3453 | 6011914 | ||

C1orf198
Genomic Map